ContactUsLinkComponent
Quick-connect coupling
  229-5123
Quick-connect coupling
  229-5123
  • Tubing int.Ø:
    3,2 mm
  • Description:
    For connecting and disconneting hoses and devices, with 1/8“ hose barb and shutoff
  • Pk:
    1
Quick-connect coupling and plug connection, made of acetal (POM) for a complete connection.
  • High mechanical stability
  • Extremely fatigue strength
  • Good electrical insulating features
  • Excellent chemical resistance
  • Both parts are with barrier