Orders LinkContactUsLinkComponent
Hydrometer
Hydrometer
Supplier:   Amarell
Hydrometer
Supplier:  Amarell
Certificates
4 Options Available Below
Product Details & Documents
General purpose hydrometers with reference temperature of 20 °C.
Certifications: DIN 12793.
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • 1,10 - 1,50 g/cm³
    0,005 g/cm³
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,00 €
    44,00 €
    1 * 1 ST
       44,00 €
  • 0,60 - 2,02 g/cm³
    0,005 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,50 €
    59,50 €
    1 * 1 ST
       59,50 €
  • 0,85 - 1,10 g/cm³
    0,005 g/cm³
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,90 €
    34,90 €
    1 * 1 ST
       34,90 €
  • 1,50 - 2 g/cm³
    0,005 g/cm³
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,00 €
    44,00 €
    1 * 1 ST
       44,00 €