Orders LinkContactUsLinkComponent
System Boxes
System Boxes
System Boxes
Certificates
System Boxes
2 Options Available Below
Product Details & Documents
Boxes made of transparent glass-clear polystyrene. Suitable for variable applications.
  • Provides immediate overview about content and quantity
  • Large sight opening
  • High stackability
  • Wide labelling pad
  • Excellent stability
Product Family Options
Product Information
  • Colour
    Dimensions
    Availability
    Price
  • Transparent
    160×100×75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
  • Transparent
    230×150×130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,70 €
    17,70 €
    1 * 1 ST
       17,70 €