Orders LinkContactUsLinkComponent
Beakers, low form, PMP
Beakers, low form, PMP
Catalog #
Beakers, low form, PMP
Catalog #
Supplier:  
Beakers, low form, PMP
Beakers, low form, PMP
Product Details & Documents
PMP, transparent.
  • Good chemical resistance
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
  • For autoclaving (121 °C) beakers with an embossed scale should be selected
With beaded rim, spout and blue printed scale.
Certifications: ISO 7056 - 10 ml/150 ml/400 ml/600 ml/3000 ml in addition to ISO 7056