Orders LinkContactUsLinkComponent
Beakers, low form, PP | Avantor
Beakers, low form, PP
Supplier:   Brand
Beakers, low form, PP
Supplier:  Brand
Certificates
Beakers, low form, PP
Beakers, low form, PP
13 Options Available Below
Product Details & Documents
PP, highly translucent.
  • Excellent chemical resistance
  • Cleaning up to max 60 °C is recommended to preserve marks and inscriptions
  • For autoclaving (+121 °C) beakers with an embossed scale should be selected
With printed blue scale, beaded rim and spout.
Certifications: ISO 7056 - 10 ml/150 ml/400 ml/600 ml/3000 ml in addition to ISO 7056
Dokumente
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Graduation Interval
    Availability
    Price
  • 10 ml
    36 mm
    30 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 22,20 €
    22,20 €
    1 * 12 ST
       22,20 €
  • 25 ml
    50 mm
    38 mm
    5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 22,20 €
    22,20 €
    1 * 12 ST
       22,20 €
  • 50 ml
    60 mm
    47 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 23,40 €
    23,40 €
    1 * 12 ST
       23,40 €
  • 100 ml
    70 mm
    55 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 27,00 €
    27,00 €
    1 * 12 ST
       27,00 €
  • 150 ml
    80 mm
    66 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 31,20 €
    31,20 €
    1 * 12 ST
       31,20 €
  • 250 ml
    95 mm
    77 mm
    25 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 17,40 €
    17,40 €
    1 * 6 ST
       17,40 €
  • 400 ml
    112 mm
    87 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 24,00 €
    24,00 €
    1 * 6 ST
       24,00 €
  • 500 ml
    118 mm
    94 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 24,90 €
    24,90 €
    1 * 6 ST
       24,90 €
  • 600 ml
    127 mm
    100 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 25,20 €
    25,20 €
    1 * 6 ST
       25,20 €
  • 1000 ml
    147 mm
    120 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 36,60 €
    36,60 €
    1 * 6 ST
       36,60 €
  • 2000 ml
    187 mm
    149 mm
    200 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 67,20 €
    67,20 €
    1 * 6 ST
       67,20 €
  • 3000 ml
    212 mm
    170 mm
    250 ml
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 88,40 €
    88,40 €
    1 * 4 ST
       88,40 €
  • 5000 ml
    247 mm
    203 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 116,00 €
    116,00 €
    1 * 4 ST
       116,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.