Order Entry
Austria
ContactUsLinkComponent
First aid case, SN-CD
First aid case, SN-CD
Catalog # 118-1226
Supplier:  SOEHNGEN
First aid case, SN-CD
Catalog # 118-1226
Supplier:  SOEHNGEN

Specifications

  • Description:
    First aid case, orange ABS plastic, with filling according to DIN 13157 (Industry norm)
  • Dimensions:
    310×130×210 mm
  • Pk:
    1

Specifications

About this item

This first aid case made of ABS plastic offers sufficient space for all of the DIN 13157 equipment.

  • Wall mounting bracket with 90 °C stop-lock
  • Can be used mobile or stationary
  • With carrying handle
  • With colour pictograms
Certifications: DIN 13157