Blotting membranes, Amersham™ Hybond™ LFP 0,2 PVDF
Supplier: Cytiva
Certificates
About this item
Amersham™ Hybond™ LFP 0.2 PVDF is a low fluorescent, hydrophobic membrane, with 0,2 µm pore size, for protein transfer. Amersham™ Hybond™ LFP is optimised for fluorescent detection in Western blotting applications and for use with the Amersham™ ECL Plex™ detection system.
- Very low background fluorescence resulting in high sensitivity
- Provides low auto-fluorescence across a wide range of excitation/emission wavelengths
- Available in popular pre-cut sizes
- Protein binding capacity >200 μg/cm²
- Compatible with a variety of blocking agents, transfer buffers, and total protein blot stains
For convenience, Western blotting sandwiches (10 sandwiches/package) are also available. Each sandwich consists of pre-cut Amersham™ Hybond™ LFP 0,2 PVDF membranes pre-assembled with 2×3 mm Chr filter papers.
Related Information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together