Orders LinkContactUsLinkComponent
Autosampler syringes
Autosampler syringes
Autosampler syringes
Certificates
Autosampler syringes
Autosampler syringes
33 Options Available Below
Product Details & Documents
For HP7673, 7683, 7693A and 6850ALS.
Gas tight
Suitable for use with Merlin Microseal™ injector.
Product Family Options
Product Information
  • Gauge
    Volume
    Length
    Ext.Ø
    Type of tip
    Type
    Availability
    Price
  • 549-0382
    Removable straight needle
    Removable straight needle
    26
    0,5 µl
    42 mm
    0,47 mm
    Cone
    0.5BR-AG-0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 160,00 €
    160,00 €
    1 * 1 ST
       160,00 €
  • 562-1002
    Removable straight needle **
    Removable straight needle **
    23
    0,5 µl
    42 mm
    0,63 mm
    Cone
    0.5BR-AG-0.63C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 160,00 €
    160,00 €
    1 * 1 ST
       160,00 €
  • 549-0391
    Removable tapered needle
    Removable tapered needle
    23-26
    0,5 µl
    42 mm
    0,63/0,47 mm
    Cone
    0.5BR-AG-0.63/0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 160,00 €
    160,00 €
    1 * 1 ST
       160,00 €
  • 549-0383
    Removable straight needle
    Removable straight needle
    23
    1 µl
    42 mm
    0,63 mm
    Cone
    1BR-AG-0.63C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 137,00 €
    137,00 €
    1 * 1 ST
       137,00 €
  • 549-0371
    Fixed straight needle
    Fixed straight needle
    26
    5 µl
    42 mm
    0,47 mm
    Cone
    5F-AG-0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,50 €
    92,50 €
    1 * 1 ST
       92,50 €
  • 562-1014
    Fixed straight needle
    Fixed straight needle
    26
    5 µl
    42 mm
    0,47 mm
    Cone
    5F-AG-0.47C
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 487,00 €
    487,00 €
    1 * 6 ST
       487,00 €
  • 562-1015
    Fixed straight needle **
    Fixed straight needle **
    23
    5 µl
    42 mm
    0,63 mm
    Cone
    5F-AG-0.63C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,50 €
    92,50 €
    1 * 1 ST
       92,50 €
  • 562-1016
    Fixed straight needle **
    Fixed straight needle **
    23
    5 µl
    42 mm
    0,63 mm
    Cone
    5F-AG-0.63C 6 pack
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 482,00 €
    482,00 €
    1 * 6 ST
       482,00 €
  • 549-0374
    Fixed tapered needle
    Fixed tapered needle
    23-26
    5 µl
    42 mm
    0,63/0,47 mm
    Cone
    5F-AG-0.63/0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,70 €
    93,70 €
    1 * 1 ST
       93,70 €
  • 568-0344
    Fixed tapered needle
    Fixed tapered needle
    23-26
    5 µl
    42 mm
    0,63/0,47 mm
    Cone
    5F-AG-0.63/0.47C 6 pack
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 473,00 €
    473,00 €
    1 * 6 ST
       473,00 €
  • 562-1034
    Fixed straight needle
    Fixed straight needle
    26
    10 µl
    42 mm
    0,47 mm
    Cone
    10F-AG-0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,80 €
    63,80 €
    1 * 1 ST
       63,80 €
  • 562-1035
    Fixed straight needle
    Fixed straight needle
    26
    10 µl
    42 mm
    0,47 mm
    Cone
    10F-AG-0.47C 6 pack
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 323,00 €
    323,00 €
    1 * 6 ST
       323,00 €
  • 549-0384
    Removable straight needle
    Removable straight needle
    26
    10 µl
    42 mm
    0,47 mm
    Cone
    10R-AG-0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,70 €
    93,70 €
    1 * 1 ST
       93,70 €
  • 549-0372
    Fixed straight needle **
    Fixed straight needle **
    23
    10 µl
    42 mm
    0,63 mm
    Cone
    10F-AG-0.63C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,20 €
    61,20 €
    1 * 1 ST
       61,20 €
  • 568-0345
    Fixed straight needle **
    Fixed straight needle **
    23
    10 µl
    42 mm
    0,63 mm
    Side hole/dome
    10F-AG-4.2/0.63H
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,10 €
    73,10 €
    1 * 1 ST
       73,10 €
  • 549-0373
    Fixed straight needle * **
    Fixed straight needle * **
    23
    10 µl
    42 mm
    0,63 mm
    Cone
    10F-AG-GT-0.63C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,60 €
    87,60 €
    1 * 1 ST
       87,60 €
  • 568-0346
    Fixed straight needle * **
    Fixed straight needle * **
    23
    10 µl
    42 mm
    0,63 mm
    Cone
    10F-AG-0.63C 25 pack
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 1 170,00 €
    1 170,00 €
    1 * 25 ST
       1 170,00 €
  • 562-1036
    Fixed straight needle * **
    Fixed straight needle * **
    23
    10 µl
    42 mm
    0,63 mm
    Cone
    10F-AG-0.63C 6 pack
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 327,00 €
    327,00 €
    1 * 6 ST
       327,00 €
  • 562-1037
    Removable straight needle **
    Removable straight needle **
    23
    10 µl
    42 mm
    0,63 mm
    Cone
    10R-AG-0.63C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,70 €
    93,70 €
    1 * 1 ST
       93,70 €
  • 568-0347
    Fixed straight needle * **
    Fixed straight needle * **
    23
    10 µl
    42 mm
    0,63 mm
    Cone
    10F-AG-GT-0.63C 25 pack
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 1 880,00 €
    1 880,00 €
    1 * 25 ST
       1 880,00 €
  • 568-0348
    Removable straight needle * **
    Removable straight needle * **
    23
    10 µl
    42 mm
    0,63 mm
    Cone
    10R-AG-GT-0.63C 10 pack
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 792,00 €
    792,00 €
    1 * 10 ST
       792,00 €
  • 549-0375
    Fixed tapered needle
    Fixed tapered needle
    23-26
    10 µl
    42 mm
    0,63/0,47 mm
    Cone
    10F-AG-0.63/0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,20 €
    61,20 €
    1 * 1 ST
       61,20 €
  • 568-0349
    Fixed tapered needle
    Fixed tapered needle
    23-26
    10 µl
    42 mm
    0,63/0,47 mm
    Cone
    10F-AG-0.63/0.47C 6 pack
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 330,00 €
    330,00 €
    1 * 6 ST
       330,00 €
  • 568-0350
    Fixed tapered needle
    Fixed tapered needle
    23-26
    10 µl
    42 mm
    0,63/0,47 mm
    Cone
    10F-AG-0.63/0.47C 25 pack
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 1 300,00 €
    1 300,00 €
    1 * 25 ST
       1 300,00 €
  • 549-0376
    Fixed tapered needle *
    Fixed tapered needle *
    23-26
    10 µl
    42 mm
    0,63/0,47 mm
    Cone
    10F-AG-GT-0.63/0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 91,50 €
    91,50 €
    1 * 1 ST
       91,50 €
  • 568-0351
    Fixed straight needle * **
    Fixed straight needle * **
    23-26
    10 µl
    42 mm
    0,63/0,47 mm
    Cone
    10F-AG-GT-0.63/0.47C
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 507,00 €
    507,00 €
    1 * 6 ST
       507,00 €
  • 549-0394
    Removable tapered needle *
    Removable tapered needle *
    23-26
    10 µl
    42 mm
    0,63/0,47 mm
    Cone
    10R-AG-GT-0.63/0.47C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,80 €
    96,80 €
    1 * 1 ST
       96,80 €
  • 568-0352
    Fixed straight needle
    Fixed straight needle
    26
    10 µl
    42 mm
    0,47 mm
    Cone
    10FX-AG-0.47 6 pack
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 486,00 €
    486,00 €
    1 * 6 ST
       486,00 €
  • 568-0353
    Fixed straight needle
    Fixed straight needle
    23
    10 µl
    42 mm
    0,63 mm
    Cone
    10FX-AG-0.63 6 pack
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 464,00 €
    464,00 €
    1 * 6 ST
       464,00 €
  • 549-0377
    Fixed straight needle *
    Fixed straight needle *
    23-26
    25 µl
    42 mm
    0,63/0,47 mm
    Cone
    25F-AG-GT-0.63/0.47
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • 549-0388
    Removable straight needle
    Removable straight needle
    23
    50 µl
    42 mm
    0,63 mm
    Cone
    50R-AG-0.63C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 127,00 €
    127,00 €
    1 * 1 ST
       127,00 €
  • 549-0378
    Fixed straight needle *
    Fixed straight needle *
    23-26
    50 µl
    42 mm
    0,63/0,47 mm
    Cone
    50F-AG-GT-0.63/0.47
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  • 549-0379
    Fixed straight needle *
    Fixed straight needle *
    23-26
    100 µl
    42 mm
    0,63/0,47 mm
    Cone
    100F-AGGT-0.63/0.47
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 175,00 €
    175,00 €
    1 * 1 ST
       175,00 €