Blotting membranes, Amersham™ Hybond™ 0.2 PVDF
: Cytiva
Amersham™ Hybond™ 0.2 PVDF is a 0,2 μm pore size hydrophobic membrane with high physical strength. This results in significant handling advantages over unsupported nitrocellulose and makes the membrane highly suitable for stripping and reprobing. The small pore size of the membrane minimises 'blow-through' and increases protein binding over a wide range of molecular weights. Ideal for use with standard colorimetric and chemiluminescent detection methods for proteins of
- High protein retention, even after harsh washing steps
- Protein binding capacity over 200 μg/cm² for easy signal detection
- Maximum capture of proteins during transfers minimises sample loss
- 0,2 μm pore size for higher surface area, resulting in better binding of low molecular weight proteins
- Available in popular pre-cut sizes
For convenience, Western blotting sandwiches (10 sandwiches/package) are also available. Each sandwich consists of pre-cut Amersham™ Hybond™ 0.2 PVDF membranes pre-assembled with 2×3 mm Chr filter papers.
Frequently Bought Together