Orders LinkContactUsLinkComponent
Bath fluids, Haake
Bath fluids, Haake
Supplier:   THERMO ELECTRON LED
Bath fluids, Haake
Certificates
Bath fluids, Haake
12 Options Available Below
Product Details & Documents
These carefully selected and proven heat transfer liquids have been designed to reduce health hazards and minimise unpleasant odours.
  • Silicone oils carry a low risk of inflammation, do not give off unpleasant odours and have a long service life
  • Low corrosive tendency
The oils are not suitable for contact with silicone tubing.
Documents
Product Family Options
Product Information
  • Type
    Temperature range
    Fire point
    Flash point
    Viscosity
    Colour
    Pk
    Availability
    Price
  • Synth 60
    –50...+45 °C
    70 °C
    59 °C
    2 cP
    Colourless
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 309,00 €
    309,00 €
    1 * 5 L
       309,00 €
  • Synth 60
    –50... +45 °C
    70 °C
    59 °C
    2 cP
    Colourless
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 569,00 €
    569,00 €
    1 * 10 L
       569,00 €
  • Sil 100
    –75...+75 °C
    >100 °C
    57 °C
    3 cP
    Colourless
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 649,00 €
    649,00 €
    1 * 5 L
       649,00 €
  • Sil 100
    –75...+75 °C
    >100 °C
    57 °C
    3 cP
    Colourless
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 1 220,00 €
    1 220,00 €
    1 * 10 L
       1 220,00 €
  • Sil 180
    –40...+200 °C
    >225 °C
    170 °C
    11 cP
    Colourless
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 383,00 €
    383,00 €
    1 * 5 L
       383,00 €
  • Sil 180
    –40...+200 °C
    >225 °C
    170 °C
    11 cP
    Colourless
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 655,00 €
    655,00 €
    1 * 10 L
       655,00 €
  • Synth 200
    +20...+210 °C
    >235 °C
    227 °C
    100 cP
    Light brown
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 582,00 €
    582,00 €
    1 * 1 ST
       582,00 €
  • Synth 200
    +20...+210 °C
    >235 °C
    227 °C
    100 cP
    Light brown
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 1 100,00 €
    1 100,00 €
    1 * 10 L
       1 100,00 €
  • Synth 260
    +50...+250 °C
    275 °C
    260 °C
    140 cP
    Yellow
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 344,00 €
    344,00 €
    1 * 5 L
       344,00 €
  • Synth 260
    +50...+ 250 °C
    275 °C
    260 °C
    140 cP
    Yellow
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 636,00 €
    636,00 €
    1 * 10 L
       636,00 €
  • Synth 260
    +50...+ 250 °C
    275 °C
    260 °C
    140 cP
    Yellow
    10 L
    Discontinued
  • Sil 300
    +80...+300 °C
    >325 °C
    300 °C
    200 cP
    Colourless
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 1 280,00 €
    1 280,00 €
    1 * 5 L
       1 280,00 €
  • Sil 300
    +80...+300 °C
    >325 °C
    300 °C
    200 cP
    Colourless
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 1 800,00 €
    1 800,00 €
    1 * 10 L
       1 800,00 €