Orders LinkContactUsLinkComponent
Multi-parameter depth probes, MPP 930 IDS
Multi-parameter depth probes, MPP 930 IDS
Supplier:   WTW
Multi-parameter depth probes, MPP 930 IDS
Supplier:  WTW
Certificates
Multi-parameter depth probes, MPP 930 IDS
Multi-parameter depth probes, MPP 930 IDS
2 Options Available Below
Product Details & Documents
These digital depth probes, with built-in barometric pressure and temperature compensation, combine dissolved oxygen, pH or ORP and conductivity measurements with accurate depth readings. They can be used with either one, two or three IDS electrodes. The multi-parameter meter Multi 3430 or Multi 3630 IDS is required for measurements.
  • No twisting, no screwing: Only a click is needed for creating a pressure, tension resistant, data proof connection to the MPP IDS
  • Optional special cable reel with sliding contacts for simultaneously winding the cable and taking readings; space for cables up to 100 m length
  • Three sensor inputs for simultaneously measuring up to three parameters, including depth
Ordering information: Sensors on request, please contact customer service.
Delivery information: Probes are supplied with built-in pressure sensor, guard, weight, and protection caps for sensor inputs and probe output. Sensors need to be ordered separately.
The kit is supplied with probe, built-in pressure sensor, guard, weight, and protection caps, carrying case and three sensors (SensoLyt®900-P, TetraCon® 925-P and FDO® 925).
Product Family Options
Depth probes
Product Information
  • Type
    Temp.
    Ø×L
    Connection
    For
    Availability
    Price
  • 665-0357
    Triple channel probe without sensor
    Triple channel probe without sensor
    MPP 930 IDS
    −5...50 °C
    70×500 mm
    1,5 m connection cable AS IDS/1.5
    Depth measurements to 100 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 050,00 €
    4 050,00 €
    1 * 1 ST
       4 050,00 €
  • 665-0358
    pH/DO/conductivity kit, incl. triple channel probe, SensoLyt® 900-P, TetraCon® 925 -P, FDO® 925-P and accessories
    pH/DO/conductivity kit, incl. triple channel probe, SensoLyt® 900-P, TetraCon® 925 -P, FDO® 925-P and accessories
    MPP 930-pH/FDO®/Cond Kit
    −5...50 °C
    70×500 mm
    1,5 m connection cable AS IDS/1.5
    Depth measurements to 100 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 470,00 €
    6 470,00 €
    1 * 1 ST
       6 470,00 €
IDS sensors
Product Information
  • Catalog #
    pH
    Temp.
    Electrolyte
    Diaphr.
    Connection
    Ø×L
    Material
    Description
    Type
    Availability
    Price
  • 2 - 12
    0...+60 °C
    Polymer
    Hole
    Plug head for AS IDS cable or wireless module
    12×120 mm
    Glass
    IDS pH electrode, pressure resistant (10 bar), SensoLyt® 900-P, wireless ready
    SensoLyt® 900-P
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 575,00 €
    575,00 €
    1 * 1 ST
       575,00 €
Product Information
  • Catalog #
    Connection
    Electrolyte
    Type
    pH
    Material
    Diaphr.
    Temp.
    Ø×L
    Description
    Availability
    Price
  • Glass
    0...+60 °C
    IDS ORP electrode, pressure resistant (10 bar), SensoLyt® ORP 900-P, wireless ready
    12×120 mm
    21
    Plug head for AS IDS cable or wireless module
    Hole
    Polymer
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 684,00 €
    684,00 €
    1 * 1 ST
       684,00 €
Cables
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-1,5, connection cable for IDS sensors with waterproof plug head
    1,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-15, connection cable for IDS sensors with waterproof plug head
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 224,00 €
    224,00 €
    1 * 1 ST
       224,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-20, connection cable for IDS sensors with waterproof plug head
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 245,00 €
    245,00 €
    1 * 1 ST
       245,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-3, connection cable for IDS sensors with waterproof plug head
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 155,00 €
    155,00 €
    1 * 1 ST
       155,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-10, connection cable for IDS sensors with waterproof plug head
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,00 €
    196,00 €
    1 * 1 ST
       196,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-100, connection cable for IDS sensors with waterproof plug head
    100 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-60, connection cable for IDS sensors with waterproof plug head
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 846,00 €
    846,00 €
    1 * 1 ST
       846,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • AS/IDS-40, connection cable for IDS sensors with waterproof plug head
    40 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 703,00 €
    703,00 €
    1 * 1 ST
       703,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • A 325/K, plastic armour, with protective basket, for CellOx® 325... and TetraCon® 325...
    A 325/K, plastic armour, with protective basket, for CellOx® 325... and TetraCon® 325...
    663-1851
    Log in for availability
    1 * 1 ST - 313,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Armoring A 925/K, with plastic protective hood, for TetraCon® 925-P, SensoLyt® 900-P, SensoLyt® ORP 900-P and FDO® 925-P
    Armoring A 925/K, with plastic protective hood, for TetraCon® 925-P, SensoLyt® 900-P, SensoLyt® ORP 900-P and FDO® 925-P
    665-0307
    Log in for availability
    1 * 1 ST - 319,00 €
Product Information
  • Catalog #
    Availability
    Price
  • BPI/IDS 900, coded protection cap for IDS sensor input of the MPP probe
    BPI/IDS 900, coded protection cap for IDS sensor input of the MPP probe
    662-1728
    Log in for availability
    1 * 1 ST - 89,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Cable reel, KT MPP/SK, with sliding contacts for connecting an AS/IDS-x cable
    Cable reel, KT MPP/SK, with sliding contacts for connecting an AS/IDS-x cable
    662-1727
    Log in for availability
    1 * 1 ST - 1 100,00 €
Product Information
  • Catalog #
    Availability
    Price
  • A 925-P/S, removable stainless steel armor for IDS field sensors, including guard
    A 925-P/S, removable stainless steel armor for IDS field sensors, including guard
    662-1726
    Log in for availability
    1 * 1 ST - 526,00 €
Product Information
  • Catalog #
    Availability
    Price
  • A 925-P/K, removable plastic armour for IDS field sensors, including guard
    A 925-P/K, removable plastic armour for IDS field sensors, including guard
    662-1725
    Log in for availability
    1 * 1 ST - 312,00 €
Product Information
  • Catalog #
    Availability
    Price
  • A 925-P, armour for protecting an IDS sensor when guard exists (A 925/K or A 925/S)
    A 925-P, armour for protecting an IDS sensor when guard exists (A 925/K or A 925/S)
    662-1724
    Log in for availability
    1 * 1 ST - 207,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement electrode holder for MPP 930 with titanium bail, included in set
    Replacement electrode holder for MPP 930 with titanium bail, included in set
    662-1723
    Log in for availability
    1 * 1 ST - 257,00 €