Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Human skeleton replica

Disarticulated human skeleton replica, 4,8 kg, 485×270×425 mm
Catalog # BINH1000025
Marketsource
Supplier:   3B Scientific
Human skeleton replica
Catalog # BINH1000025
Supplier:  3B Scientific
Marketsource
Human skeleton replica
Human skeleton replica

Disarticulated human skeleton replica, 4,8 kg, 485×270×425 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

The replica of the human dis-articulated skeleton includes one hand and foot on wire, and one hand and foot loosely articulated.

  • Skull's calvarium sectioned for a closeup view, and the jaw is hinged
  • Sternum's intracostal cartilage is attached
  • Every other bone is life-sized and separated

This disarticulated skeleton is great for any study of the anatomy of the human skeleton.

 : Supplied in a sturdy partitioned cardboard storage box.

Specifications
  • Weight:
    4,8 kg
  • Description:
    Disarticulated human skeleton replica
  • W×D×H:
    485×270×425 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.