Labels for petri dishes, PetriStickers™
Supplier: DIVERSIFIED BIOTECH
Certificates
About this item
PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably.
- Temperature resistant from +4 to +42° C
- Water-resistant ink
Simply remove the label's protective backing and apply it to the back of a petri dish. After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.
Frequently Bought Together