Order Entry
Austria
ContactUsLinkComponent
Screw caps with aperture, BOLA
Screw caps with aperture, BOLA
Catalog # BOHLH999-45
Supplier:  Bohlender
undefined
Screw caps with aperture, BOLA
Catalog # BOHLH999-45
Supplier:  Bohlender

Specifications

  • Thread:
    GL 45
  • Colour:
    Green
  • Pk:
    1

Specifications

About this item

Screw caps, made of PP, with handy knurl.

  • Diameter of aperture: 34 mm
  • Good chemical resistance
  • Temperature resistance: from –20 to +110 °C