Caps for storage tubes, 2D barcoded, Matrix®
Lieferant: THERMO MATRIX TECHNOLOGIES
Zertifikate
Über diesen Artikel
Simplify sample storage with these color-coded caps. Made from Virgin class VI medical grade PP with virgin class VI medical grade silicone O-ring.
- Free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
- Gasketed caps ensure critical sample protection over the storage lifetime
- Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
- ScrewTop caps are compatible with storage down to −176 °C
Spezifikationen
- Catalog No:
- 479-2050
- 479-2051
- 479-2048
- 479-2049
- 479-1338
- 479-2046
- 479-2047
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Spezifikationen
Frequently Bought Together