ContactUsLinkComponent
Glove dispensers
Glove dispensers
Artikelnr. SCURGD-4PETG
Lieferant:  S-CURVE
undefined
Glove dispensers
Artikelnr. SCURGD-4PETG
Lieferant:  S-CURVE
Spezifikationen
  • Colour:
    Clear
  • Description:
    Gloves four-compartments dispenser with lid
  • W×D×H:
    508×292×305 mm
  • Pk:
    1
Spezifikationen
Über diesen Artikel
Polyethylene terephthalate glycol (PETG) dispenser with separate compartments and lids designed for gloves.
  • Wall mounting or free standing
  • Heavy duty wall bracket included
  • Flame treated edges for smoothness
Suitable for applications in cleanrooms.