Sample containers, with screw cap
Supplier: DELTALAB
Jump to:
About this item
Container made of PS with PE screw cap.
- Good chemical resistance
- Aseptic or sterile available
- With high safety closure biflex caps
Recommended volume: 50 ml
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
3 Options Available Below
Product Details & Documents
Container made of PS with PE screw cap.
- Good chemical resistance
- Aseptic or sterile available
- With high safety closure biflex caps
Recommended volume: 50 ml
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
- CapacityHeightColourCap colourØ top/Ø bottomVersionAvailabilityPrice
- DELT309502Sample container
- DELT309552Sample container, with spoonSample container, with spoon60 ml66 mmClearRed39,3/44,2 mmasepticLog in for availability1 * 600 ST446,00 €
- DELT309526Sample container, individually wrappedSample container, individually wrapped60 ml66 mmClearRed39,3/44,2 mmsterileLog in for availability1 * 750 ST392,00 €