Orders LinkContactUsLinkComponent
Microplates, 384-well, non binding | Avantor
Microplates, 384-well, non binding
   Sustainable Options Available
Supplier:   Greiner Bio-One
Microplates, 384-well, non binding
Supplier:  Greiner Bio-One
   Sustainable Options Available
Certificates
7 Options Available Below
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
All plates are free from RNase, DNase, and pyrogens, and are also free of DNA polymerase inhibitors.
Specifications for Product Family
Specifications
  • Catalog No:
  • 738-0175
  • GREI781901_40
  • 738-0177
  • 738-0179
  • 738-0176
  • 738-0178
  • GREI784904_40
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • 738-0175
    384-well microplate, non binding, flat bottom
    384-well microplate, non binding, flat bottom
    Black
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 367,00 €
    367,00 €
    1 * 40 ST
       367,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0175
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • GREI781901_40
    384-well microplate, non binding, flat bottom
    384-well microplate, non binding, flat bottom
    Clear
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 354,00 €
    354,00 €
    1 * 40 ST
       354,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI781901_40
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0177
    384-well microplate, non binding, flat bottom
    384-well microplate, non binding, flat bottom
    White
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 339,00 €
    339,00 €
    1 * 40 ST
       339,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0177
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0179
    384-well microplate, non binding, flat bottom
    384-well microplate, non binding, flat bottom
    Black
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 494,00 €
    494,00 €
    1 * 40 ST
       494,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0179
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0176
    384-well microplate, non binding, flat μClear® bottom
    384-well microplate, non binding, flat μClear® bottom
    White/clear
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 510,00 €
    510,00 €
    1 * 40 ST
       510,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0176
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • 738-0178
    384-well microplate, non binding, flat μClear® bottom
    384-well microplate, non binding, flat μClear® bottom
    Black/clear
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 485,00 €
    485,00 €
    1 * 40 ST
       485,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    738-0178
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
  • GREI784904_40
    384-well microplate, Small Volume™, HiBase, non binding, flat bottom
    384-well microplate, Small Volume™, HiBase, non binding, flat bottom
    White
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 854,00 €
    854,00 €
    1 * 40 ST
       854,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    GREI784904_40
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (241002); ISO 50001 (241002)
      Low Manufacturing Impact
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.