ContactUsLinkComponent
Cell culture plates, 384-well, Small Volume™, CELLSTAR®
Cell culture plates, 384-well, Small Volume™, CELLSTAR®
Catalog # 392-0371
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
undefined
Cell culture plates, 384-well, Small Volume™, CELLSTAR®
Catalog # 392-0371
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Specifications
  • Recommended working volume:
    4 - 25 µl
  • Colour:
    White
  • Description:
    Solid bottom, HiBase, Low Volume™, with lid
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Culture area:
    0,27 cm²
  • Pk:
    32
  • Well volume:
    30 µl
Specifications
About this item
PS, flat bottom, round wells, TC treated. These 384-well Low Volume™ microplates offer the same standardised footprint and height as regular polystyrene plates. This enables a direct change from larger volumes into the small volume range or even a combination of both plates.
  • Raised rims on each well minimise cross contamination
  • Small volume allows reagent savings