ContactUsLinkComponent
Tubing holder for beakers, SP Bel-Art
Tubing holder for beakers, SP Bel-Art
Catalog # 229-0001
undefined
Tubing holder for beakers, SP Bel-Art
Catalog # 229-0001
Specifications
  • Spanning width:
    9,5 mm
  • Colour:
    Transparent
  • Length:
    51 mm
  • Pk:
    6
Specifications
About this item
This PP clip holds flexible tubing steady and fits securely onto the edge of beakers or pitchers up to ¼" thick.
  • 5,1 cm (2") long arm has a hole to accept tubing up to 9,5 mm (⅜") Ext. Ø
  • Autoclavable