Orders LinkContactUsLinkComponent
Specific gravity cups, calibrated
Specific gravity cups, calibrated
Supplier:   Industrial Physics
Specific gravity cups, calibrated
Supplier:  Industrial Physics
Certificates
Specific gravity cups, calibrated
Specific gravity cups, calibrated
4 Options Available Below
Product Details & Documents
Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
  • Density is defined as weight per unit volume at a specified temperature
  • Available in anodised aluminium or stainless steel
Certifications: According to ISO 2811, DIN 53 217 and ASTM D 1475.
Delivery information: Supplied with a calibration certificate.
Documents
Product Family Options
Product Information
  • Volume (nom.)
    Availability
    Price
  • SIMEVF2098
    Anodised aluminium
    Anodised aluminium
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 337,00 €
    337,00 €
    1 * 1 ST
       337,00 €
  • SIMEVF2097
    Anodised aluminium
    Anodised aluminium
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 321,00 €
    321,00 €
    1 * 1 ST
       321,00 €
  • SIMEVF2100
    Stainless steel
    Stainless steel
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 395,00 €
    395,00 €
    1 * 1 ST
       395,00 €
  • 614-0715
    Stainless steel
    Stainless steel
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 395,00 €
    395,00 €
    1 * 1 ST
       395,00 €