Orders LinkContactUsLinkComponent
Test sets
Test sets
Supplier:   Dräger
Test sets
Supplier:  Dräger
Certificates
Test sets
7 Options Available Below
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifications for Product Family
Specifications
  • Catalog No:
  • 151-0176
  • 151-0177
  • 151-0179
  • DRAE8103170
  • DRAE8103380
  • DRAE8103410
  • 151-0116
  • Model:
  • Test set I for inorganic gases
  • Test set II for inorganic gases
  • Test set III for organic vapours
  • Test set for conductive compounds
  • Test sets for fumes
  • Test set I for fumes
  • Measurement time:
  • 40 s
  • 40 s
  • 120 s
  • 120 s
  • 240 s
  • 180 s
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 151-0176
    Test set I for inorganic gases
    Test set I for inorganic gases
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,50 €
    84,50 €
    1 * 1 ST
       84,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    151-0176
    Model
    Test set I for inorganic gases
    Measurement time
    40 s
  • 151-0177
    Test set II for inorganic gases
    Test set II for inorganic gases
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,50 €
    84,50 €
    1 * 1 ST
       84,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    151-0177
    Model
    Test set II for inorganic gases
    Measurement time
    40 s
  • 151-0179
    Test set III for organic vapours
    Test set III for organic vapours
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,50 €
    84,50 €
    1 * 1 ST
       84,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    151-0179
    Model
    Test set III for organic vapours
    Measurement time
    120 s
  • DRAE8103170
    Test set for conductive compounds
    Test set for conductive compounds
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    DRAE8103170
    Model
    Test set for conductive compounds
    Measurement time
    120 s
  • DRAE8103380
    Test sets for fumes
    Test sets for fumes
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 173,00 €
    173,00 €
    1 * 2 ST
       173,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    DRAE8103380
    Model
    Test sets for fumes
    Measurement time
    240 s
  • DRAE8103410
    Test set I for fumes
    Test set I for fumes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,75 €
    84,75 €
    1 * 1 ST
       84,75 €
    Specifications:
    More
    Specifications
    Cat. No.
    DRAE8103410
    Model
    Test set I for fumes
    Measurement time
    180 s
  • 151-0116
    Adapter for simultaneous test via connection to a bellows pump
    Adapter for simultaneous test via connection to a bellows pump
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,75 €
    60,75 €
    1 * 1 ST
       60,75 €
    Specifications:
    More
    Specifications
    Cat. No.
    151-0116
    Model
    Measurement time