Orders LinkContactUsLinkComponent
Dropping bottles, APONORM®
Dropping bottles, APONORM®
Supplier:   MUELLER KREMPEL
Dropping bottles, APONORM®
Supplier:  MUELLER KREMPEL
Certificates
Dropping bottles, APONORM®
Dropping bottles, APONORM®
5 Options Available Below
Product Details & Documents
Dropper bottles made of amber glass.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substances
Thread according to DIN-18 and with dust protection plug, germ-free and hygienic shrink wrap packaging.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 10 ml
    65 mm
    24,5 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 19,20 €
    19,20 €
    1 * 20 ST
       19,20 €
  • 20 ml
    69 mm
    27,5 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 21,50 €
    21,50 €
    1 * 20 ST
       21,50 €
  • 30 ml
    79 mm
    33,5 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 21,90 €
    21,90 €
    1 * 20 ST
       21,90 €
  • 50 ml
    96 mm
    39,5 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 25,10 €
    25,10 €
    1 * 20 ST
       25,10 €
  • 100 ml
    111 mm
    51,0 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 33,30 €
    33,30 €
    1 * 20 ST
       33,30 €
Accessories
Product Information
  • Catalog #
    For bottles
    Description
    Cap colour
    Availability
    Price
  • 20 ml
    Pipette mount
    White/blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,00 €
    16,00 €
    1 * 10 ST
       16,00 €
Product Information
  • Catalog #
    For bottles
    Description
    Cap colour
    Availability
    Price
  • 30 ml
    Pipette mount
    White/blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,00 €
    16,00 €
    1 * 10 ST
       16,00 €
Product Information
  • Catalog #
    For bottles
    Description
    Cap colour
    Availability
    Price
  • 10 ml
    Pipette mount
    White/blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,00 €
    16,00 €
    1 * 10 ST
       16,00 €
Product Information
  • Catalog #
    For bottles
    Description
    Cap colour
    Availability
    Price
  • 50 ml
    Pipette mount
    White/blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,00 €
    16,00 €
    1 * 10 ST
       16,00 €
Product Information
  • Catalog #
    For bottles
    Description
    Cap colour
    Availability
    Price
  • 100 ml
    Pipette mount
    White/blue
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,00 €
    16,00 €
    1 * 10 ST
       16,00 €