ContactUsLinkComponent
Brushes, swivel-head, for flasks
Brushes, swivel-head, for flasks
Catalog # 149-1210
undefined
Brushes, swivel-head, for flasks
Catalog # 149-1210
Specifications
  • Handle material:
    Wood
  • Description:
    Brush with wooden handle, swivel-head
  • Total length:
    420 mm
  • Pk:
    1
Specifications
About this item
For intensive and gentle cleaning. Fits flasks from 500 to 5000 ml with SGJ/NS 29 openings.