ContactUsLinkComponent
Urine collection containers, with self-adhesive lid
Urine collection containers, with self-adhesive lid
Catalog # BOET06.073.1400
Supplier:  BOETTGER PAUL
undefined
Urine collection containers, with self-adhesive lid
Catalog # BOET06.073.1400
Supplier:  BOETTGER PAUL
Specifications
  • Capacity:
    100 ml
  • Description:
    Urine cup with printed self-adhesive lid, PP
  • Pk:
    1.000
Specifications
About this item
PP, translucent.
  • Resistant to pressure and shatterproof, graduated
  • Tight sealing lid, no leakage, no inconvenient smells
  • Lids can be easily and clearly written on
  • Content can be poured out easily after perforation
Ideal for collection and storage of urine samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904