Order Entry
Austria
Orders LinkContactUsLinkComponent
Vials for cell counters

Vials for cell counters

Vials for cell counters (Coulter, Hycell), 25,00 ml, 32×60 mm, Yes, LDPE
Catalog # BOET02.901.2100
Supplier:   BOETTGER PAUL
Vials for cell counters
Catalog # BOET02.901.2100
Supplier:  BOETTGER PAUL
Vials for cell counters
Vials for cell counters

Vials for cell counters (Coulter, Hycell), 25,00 ml, 32×60 mm, Yes, LDPE

Price
45,40 €
Log in for availability
1 * 250 ST
Product Details & Documents

Optically clear vials with parallel surfaces, free of dust or particles.

  • Covers fitted of optimal density
  • Reliable high quality-level
Specifications
  • Cap included:
    Yes
  • Cap material:
    LDPE
  • Ø×H:
    32×60 mm
  • Capacity:
    25,00 ml
  • Description:
    Vials for cell counters (Coulter, Hycell)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.