Cold traps, Type KF
About this item
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
- Easy to clean
- No stand material for holding the cold trap
- Protective casing of Dewar flask made of blue coated metal or aluminium
7 Options Available Below
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
- Easy to clean
- No stand material for holding the cold trap
- Protective casing of Dewar flask made of blue coated metal or aluminium
Product Family Options
Product Information
- AvailabilityPrice
- 478-4422Cold trap, KF 29-K, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent capCold trap, KF 29-K, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent capLog in for availability1 * 1 ST640,00 €
- KGWA1735Cold trap, KF 29-OK, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent capCold trap, KF 29-OK, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent capLog in for availability1 * 1 ST673,00 €
- KGWA17371Cold trap, KF 29-OK-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacityCold trap, KF 29-OK-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacityLog in for availability1 * 1 ST741,00 €
- KGWA17372Cold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent capCold trap, KF 29-GL-A, borosilicate glass 3,3, with GL 32 glass thread and screw-on cap, 150 ml condensate capacity and 1000 ml coolent capLog in for availability1 * 1 ST698,00 €
- KGWA17377Cold trap S 29 GL-A, borosilicate glass 3,3, with threaded glass joint GL 18 and PTFE olive, 150 ml condensate capacityCold trap S 29 GL-A, borosilicate glass 3,3, with threaded glass joint GL 18 and PTFE olive, 150 ml condensate capacityLog in for availability1 * 1 ST367,00 €
- KGWA1740Cold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent capCold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent capLog in for availability1 * 1 ST630,00 €
- KGWA17402Cold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mmCold trap, KF 29-GL-Z, borosilicate glass 3,3, with conical ground joint NS45/40, 150 ml condensate capacity, 1000 ml coolent cap and length of 200 mmLog in for availability1 * 1 ST812,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Plastic ring for cold trap, type KF
Product Information
- Catalog #AvailabilityPrice
- Cooling finger S 29-GL