Order Entry
Austria
Orders LinkContactUsLinkComponent
H-frame COMPLETE

H-frame COMPLETE

H-frame, 6,5 kg, 500×250×800 mm
Catalog # BOCH5641
Supplier:   BOCHEM
H-frame COMPLETE
Catalog # BOCH5641
Supplier:  BOCHEM
H-frame COMPLETE
H-frame COMPLETE

H-frame, 6,5 kg, 500×250×800 mm

Price
313,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Cast iron, green varnished.

  • Incl. aluminium rods (Diameter 12 mm)
  • Incl. bossheads
Specifications
  • Description:
    H-frame
  • W×D×H:
    500×250×800 mm
  • Weight:
    6,5 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.