ContactUsLinkComponent
Critical care syringes, BD Vacutainer®
Critical care syringes, BD Vacutainer®
Catalog # BDAM364356
Supplier:  BD Medical
undefined
Critical care syringes, BD Vacutainer®
Catalog # BDAM364356
Supplier:  BD Medical
Specifications
  • Capacity:
    1,0 ml
  • Description:
    BD A-Line™ critical care syringes, without needle, slip tip, with 30 units of spray dried heparin
  • Pk:
    100
Specifications
About this item
Critical care syringes can be used to measure arterial blood gas, ionised calcium, ionised magnesium, as well as perform critical care tests.
  • Permits aspiration
Syringes contain calcium balanced lithium heparin spray dried on the sides of the syringe.