Orders LinkContactUsLinkComponent
Multi-position heaters, behrotest®
 :   BEHR
Multi-position heaters, behrotest®
 :  BEHR
Multi-position heaters, behrotest®
Multi-position heaters, behrotest®
Multi-position hotplates with protective metal grills. Maximum plate temperature is 400 °C.
  • Individually adjustable heating controls and indicator lights
Product Family Options
  • Description
    Heating capacity
    Type
    W×D×H
  • Four positions for flasks, HB 4 with controllable heating points, power 1440 W, with Ø 94 mm and weight ~15,1 kg
    1440 W
    With 4 plates
    530×320×740 mm
    1 * 1 ST
    1 * 1 ST - 1 210,29 €
    1 210,29 €
    1 * 1 ST
       1 210,29 €
  • Six positions for flasks, HB 6 with controllable heating points, power 2160 W, with Ø 94 mm and weight ~19,8 kg
    2160 W
    With 6 plates
    760×320×740 mm
    1 * 1 ST
    1 * 1 ST - 1 672,71 €
    1 672,71 €
    1 * 1 ST
       1 672,71 €
Accessories
  •  
    Description
    For
  • BEHRB00217700
    Brackets for HB 6, with 6 support rods
    HB 6
    1 * 1 ST - 366,58 €