Order Entry
Austria
ContactUsLinkComponent
Bossheads, TECHNO
Bossheads, TECHNO
Catalog # BOCH5670
Supplier:  BOCHEM
undefined
Bossheads, TECHNO
Catalog # BOCH5670
Supplier:  BOCHEM

Specifications

  • Material:
    Aluminium
  • Pk:
    1
  • For rod Ø:
    5 - 22 mm

Specifications

About this item

These bossheads are ideal for supporting overhead stirrers.

  • Angle: 90°
  • Thread: M8