Adenosine-5'-triphosphate disodium salt (ATP disodium salt)
Supplier: Cayman Chemical
Certificates
About this item
Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt
CAS Number:987-65-5
:MFCD00080339
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃
:551.14 g/mol
EINECS:213-579-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together