Ponceau S
Supplier: Cayman Chemical
Jump to:
About this item
Synonyms: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
- CAS Number:6226-79-5
- Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
- MDL Number:MFCD00003892
- Molecular Weight:760.56 g/mol
- Storage Temperature:Ambient
- EINECS:228-319-2
3 Options Available Below
Product Details & Documents
Synonyms: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
- CAS Number:6226-79-5
- Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
- MDL Number:MFCD00003892
- Molecular Weight:760.56 g/mol
- Storage Temperature:Ambient
- EINECS:228-319-2



