Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477