Sodium chloride 25% in aqueous solution
Catalog # SIAL73421-5L-F
Supplier: Merck
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
Specifications
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
Density:1…1.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477