Order Entry
Austria
ContactUsLinkComponent
Ammonium cerium(IV) nitrate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
Irritant
Ammonium cerium(IV) nitrate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
Catalog # CMPL3000100
CAS Number:  
Irritant
Ammonium cerium(IV) nitrate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
Catalog # CMPL3000100
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :548.22 g/mol
EINECS:240-827-6
 :GHS07
CAS Number:16774-21-3
Formula:(NH₄)₂Ce(NO₃)₆
MDL Number:MFCD00151121