Order Entry
Austria
Orders LinkContactUsLinkComponent
Cycloheximide

Cycloheximide

Toxic
Health
Environment
Supplier:   Cayman Chemical
Cycloheximide
Supplier:  Cayman Chemical
Certificates
Cycloheximide

2 Options Available Below

Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 43,00 €
    43,00 €
    1 * 100 mg
       43,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 85,00 €
    85,00 €
    1 * 1 g
       85,00 €