Order Entry
Austria
ContactUsLinkComponent
Ethidium bromide 0.625 mg/ml in aqueous solution
Health
Ethidium bromide 0.625 mg/ml in aqueous solution
Catalog # J62029.AA
CAS Number:  
Health
Ethidium bromide 0.625 mg/ml in aqueous solution
Catalog # J62029.AA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
Merck Index:14,4731
ADR:6.1,III
 :394.31 g/mol
EINECS:214-984-6
UN:2810
 :GHS08
Storage Temperature:Ambient
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724