4-(Difluoromethoxy)iodobenzene
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: 1-(Difluoromethoxy)-4-iodobenzene
CAS Number:128140-82-9
:
Formula:C₇H₅F₂IO
MDL Number:MFCD00085008
:270.0153 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together