Order Entry
Austria
ContactUsLinkComponent
2-(Methylsulphonyl)aniline hydrochloride 95%
Irritant
2-(Methylsulphonyl)aniline hydrochloride 95%
Catalog # ACRO424840025
CAS Number:  
undefined
Irritant
2-(Methylsulphonyl)aniline hydrochloride 95%
Catalog # ACRO424840025
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 g
  • Seq:
    3122

Specifications

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥94 %

Specification Test Results

About this item

 :
Melting Pt:139…141 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:205985-95-1
Formula:C₇H₁₀ClNO₂S
MDL Number:MFCD00216484