
3-Sulphobenzoic acid monosodium salt, Sigma-Aldrich®
Catalog # SIAL375411-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:224.17 g/mol
:M-Clarity™ quality level = MQ200
EINECS:241-602-5
:GHS07
CAS Number:17625-03-5
Formula:C₇H₅NaO₅S
MDL Number:MFCD00066378