
3-Sulphobenzoic acid monosodium salt ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17625-03-5
:
Formula:C₇H₅NaO₅S
MDL Number:MFCD00066378
:224.17 g/mol
EINECS:241-602-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together