Order Entry
Austria
ContactUsLinkComponent
 
Ferrozine mono-sodium salt hydrate indicator, Sigma-Aldrich®
 
undefined
Ferrozine mono-sodium salt hydrate indicator, Sigma-Aldrich®

 

  • Appearance (Colour):
    Yellow to Yellow-Green
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Yellow to Yellow-Green
  • Solubility (Turbidity)
    50 mg/ml, H2O:
    Clear to Slightly Hazy
  • Water (by Karl Fischer):
    ≤8 %
  • Sodium (Na) Anhydrous
    By ICP-atomic emission:
    4.0 - 5.9 %
  • Suitability
    For Determination of Fe++:
    Suitable
  • Recommended Retest Period:
    4 years

 

Frequently Bought Together