
Victoria blue B, pure
:2580-56-5
:
:C₃₃H₃₂ClN₃
:MFCD00011878
:506.803 g/mol
:206 °C
:219-943-6
:GHS07
:Ambient
- Appearance:Dark brown Glistening crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Loss on drying:≤5 % (60°C)
- UV Lambda max:599 to 605 nm (in ethanol)
Frequently Bought Together