1,1-Dimethylbiguanide hydrochloride, Sigma-Aldrich®
Catalog # SIALD150959-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:165.625 g/mol
:M-Clarity™ quality level = MQ200
EINECS:214-230-6
CAS Number:1115-70-4
Formula:C₄H₁₂ClN₅
MDL Number:MFCD00012582