ContactUsLinkComponent
Lithium aluminum hydride 4.0 M in diethyl ether, AcroSeal®
Irritant
Corrosive
Flammable
Lithium aluminum hydride 4.0 M in diethyl ether, AcroSeal®
Certificates
undefined
Irritant
Corrosive
Flammable
Lithium aluminum hydride 4.0 M in diethyl ether, AcroSeal®
Certificates
Specification Test Results
  • Appearance:
    Colorless Solution (May develop some turbidity or precipitate)
  • Titration Iodometric:
    18.0 to 22.0 % (LiAlH4)