Orders LinkContactUsLinkComponent
Lab markers
Lab markers
Lab markers
Supplier:  MICRONOVA
Certificates
Lab markers
2 Options Available Below
Product Details & Documents
LabMarkers feature permanent ink that marks on cold or wet surfaces, plastic bags or disposable labware. Marks will not smear on reusable glass, metal or porcelain labware but can be scrubbed off.
  • Fine tip, black ink
  • Permanent on most surfaces
  • Fast drying
  • Available gamma irradiated
Materials of construction: High-density poly barrel. Permanent, quick drying, alcohol- resistant black ink.
Chemical compatibility: Holds up to wipedown with IPA, bleach, and phenols. Integrity of printed barrel will be compromised with thinners and strong solvents.
Caution: Markers cannot be used with Ethanol or other alcohols.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • 811-0177
    LabMarker, fine tip
    LabMarker, fine tip
    Black
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 98,10 €
    98,10 €
    1 * 10 ST
       98,10 €
  • 811-0178
    LabMarker, fine tip, irradiated
    LabMarker, fine tip, irradiated
    Black
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 123,00 €
    123,00 €
    1 * 10 ST
       123,00 €