Order Entry
Austria
ContactUsLinkComponent
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution indicator for waste water analysis, Supelco®
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution indicator for waste water analysis, Supelco®
Supplier:  Merck
Certificates
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution indicator for waste water analysis, Supelco®
Supplier:  Merck

Specification Test Results

  • Spec. Absorptivity A 1%/1cm (λmax; 0.2 %; buffer pH 4.0):
    2.4 - 2.8
  • pH-value (20 °C):
    3.5 - 4.5
  • Application test:
    conforms

Specification Test Results

Related Information

Frequently Bought Together