Order Entry
Austria
ContactUsLinkComponent
VWR® pHenomenal® MU 6100 H Multi-Parameter Meter (pH/Conductivity/DO), Handheld
VWR® pHenomenal® MU 6100 H Multi-Parameter Meter (pH/Conductivity/DO), Handheld
Supplier:  VWR Collection
Certificates
undefined
VWR® pHenomenal® MU 6100 H Multi-Parameter Meter (pH/Conductivity/DO), Handheld
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 665-0312
  • 665-0311
  • Model:
  • MU 6100 H
  • MU 6100 H
  • pH range:
  • −2,000...+20,000
  • −2,000...+20,000
  • pH resolution:
  • 0,1/0,01/0,001 (selectable)
  • 0,1/0,01/0,001 (selectable)
  • pH accuracy:
  • ±0,005 + 1 digit
  • ±0,005 + 1 digit
  • pH calibration:
  • Up to 5 points
  • Up to 5 points
  • Automatic buffer recognition:
  • TITRINORM® buffers at 25 °C: 4,00/6,98/9,94
    NIST/DIN 19266 (1,68/4,00/6,86/9,18/12,54)
    Technical buffers at 25 °C: 4,00/7,00/10,00
  • TITRINORM® buffers at 25 °C: 4,00/6,98/9,94
    NIST/DIN 19266 (1,68/4,00/6,86/9,18/12,54)
    Technical buffers at 25 °C: 4,00/7,00/10,00
  • mV range:
  • ±1200,0/±2500
  • ±1200,0/±2500
  • mV resolution:
  • 0,1/1
  • 0,1/1
  • mV accuracy:
  • ±0,3/±1
  • ±0,3/±1
  • Conductivity range:
  • 0,001 µS/cm - 1000 mS/cm
  • 0,001 µS/cm - 1000 mS/cm
  • Conductivity resolution:
  • 0,1 µS/cm
  • 0,1 µS/cm
  • Conductivity accuracy:
  • ±0,5% of measured value
  • ±0,5% of measured value
  • Conductivity cell constant:
  • Depending on probe
  • Depending on probe
  • Conductivity calibration:
  • 1 point
  • 1 point
  • Resistivity range:
  • 1 Ω/cm - 999 MΩ/cm
  • 1 Ω/cm - 999 MΩ/cm
  • Resistivity accuracy:
  • ±0,5% of measured value
  • ±0,5% of measured value
  • Salinity range:
  • 0,00 - 70 ppt
  • 0,00 - 70 ppt
  • TDS range:
  • 0 - 1999 mg/L / 2,00 - 19,99 g/L / 20,0 - 199,9 g/L
  • 0 - 1999 mg/L / 2,00 - 19,99 g/L / 20,0 - 199,9 g/L
  • TDS resolution:
  • 1 mg/L / 0,01 g/L / 0,1 g/L
  • 1 mg/L / 0,01 g/L / 0,1 g/L
  • DO concentration:
  • 0,00 - 20,00 / 0,0 - 90,0 mg/L
  • 0,00 - 20,00 / 0,0 - 90,0 mg/L
  • DO concentration resolution:
  • 0,1%; 0,01 mg/L
  • 0,1%; 0,01 mg/L
  • DO saturation:
  • 0,0 - 200,0; 0 - 600 %
  • 0,0 - 200,0; 0 - 600 %
  • DO accuracy:
  • ±0,5% of value (between 5...30 °C)
  • ±0,5% of value (between 5...30 °C)
  • DO calibration:
  • Water saturated air
  • Water saturated air
  • Temperature range:
  • Auto: −5,0...+105,0 °C; manual: −25...+130 °C
  • Auto: −5,0...+105,0 °C; manual: −25...+130 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,2 °C
  • ±0,2 °C
  • Temperature compensation:
  • Automatic/manual
  • Automatic/manual
  • Display:
  • Graphic LCD with LED backlight
  • Graphic LCD with LED backlight
  • Power supply:
  • 4× AA batteries
  • 4× AA batteries
  • Compliance:
  • IP 67/GLP
  • IP 67/GLP
  • Data logging - log function:
  • Selectable between 1 s and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
  • Selectable between 1 s and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
  • Data logging capacity:
  • 5000 data sets
  • 5000 data sets
  • Connections:
  • BNC for pH/banana for temperature/8-pin for conductivity and DO
  • BNC for pH/banana for temperature/8-pin for conductivity and DO
  • Outputs:
  • Waterproof USB connection (slave, bidirectional)
  • Waterproof USB connection (slave, bidirectional)

Specifications

Related Information

pH electrode

Product Information

Frequently Bought Together