ContactUsLinkComponent
Strips of 8 PCR tubes with attached flat cap
Strips of 8 PCR tubes with attached flat cap
Catalog # DELT4096.2N
Supplier:  DELTALAB
Strips of 8 PCR tubes with attached flat cap
Catalog # DELT4096.2N
Supplier:  DELTALAB
Specifications
  • Packed:
    125 strips/case
  • Capacity:
    0,2 ml
  • Colour:
    Natural
  • Description:
    Strips of 8 PCR tubes, with individually attached flat caps
  • Pk:
    125
Specifications
About this item
Made of PP.
  • RNAse, DNAse and pyrogen free
  • Excellent chemical resistance
  • Excellent temperature resistance
Every cap embodies a shield in order to prevent contamination when being opened. Positive sealing will fully protect the contents from evaporation during the whole thermal cycle. These strips can be cut to any length while each tube has its own attached cap.